siRNA Details
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 21 | 3126 | ||
NM_001288953.1 | NP_001275882.1 | 21 | 3045 | |||
NM_001288955.1 | NP_001275884.1 | 19 | 2896 | |||
NM_020458.3 | NP_065191.2 | 20 | 3054 | |||
XM_005264439.3 | XP_005264496.1 | 21 | 3207 | |||
XM_011532998.2 | XP_011531300.1 | 22 | 3272 | |||
XM_011533000.2 | XP_011531302.1 | 17 | 2129 | |||
XM_011533001.2 | XP_011531303.1 | 13 | 1919 | |||
XM_017004524.1 | XP_016860013.1 | 19 | 2881 | |||
XM_017004525.1 | XP_016860014.1 | 20 | 2539 | |||
XM_017004526.1 | XP_016860015.1 | 17 | 2749 | |||
XM_017004527.1 | XP_016860016.1 | 20 | 3149 | |||
XM_017004528.1 | XP_016860017.1 | 21 | 3214 | |||
GenBank mRNA | AB032966.2 | - | 21 | 2634 | ||
AK056464.1 | - | 21 | 3017 | |||
AL834383.1 | - | 20 | 3054 | |||
BC001978.1 | - | 11 | 1463 | |||
BC027457.2 | - | 3 | 616 | |||
BC065554.1 | - | 17 | 2172 | |||
BC082965.1 | - | 12 | 1793 | |||
BC111487.1 | - | 21 | 3014 | 6041916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
