siRNA Details
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 21 | 2845 | ||
NM_001288953.1 | NP_001275882.1 | 21 | 2764 | |||
NM_001288955.1 | NP_001275884.1 | 19 | 2615 | |||
NM_020458.3 | NP_065191.2 | 20 | 2773 | |||
XM_005264439.3 | XP_005264496.1 | 21 | 2926 | |||
XM_011532998.2 | XP_011531300.1 | 22 | 2991 | |||
XM_011533000.2 | XP_011531302.1 | 17 | 1848 | |||
XM_011533001.2 | XP_011531303.1 | 13 | 1638 | |||
XM_017004524.1 | XP_016860013.1 | 19 | 2600 | |||
XM_017004525.1 | XP_016860014.1 | 20 | 2258 | |||
XM_017004526.1 | XP_016860015.1 | 17 | 2468 | |||
XM_017004527.1 | XP_016860016.1 | 20 | 2868 | |||
XM_017004528.1 | XP_016860017.1 | 21 | 2933 | |||
GenBank mRNA | AB032966.2 | - | 21 | 2353 | ||
AK056464.1 | - | 21 | 2736 | |||
AL834383.1 | - | 20 | 2773 | |||
BC001978.1 | - | 11 | 1182 | |||
BC027457.2 | - | 3 | 335 | |||
BC065554.1 | - | 17 | 1891 | |||
BC082965.1 | - | 12 | 1512 | |||
BC111487.1 | - | 21 | 2733 | 6041916 | ||
BC114365.2 | - | 19 | 2246 | 40038218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
