siRNA Details
Gene Symbol: DDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164232.1 | NP_001157704.1 | 18 | 4239 | ||
NM_015214.2 | NP_056029.2 | 18 | 4416 | |||
XM_005273454.2 | XP_005273511.1 | 18 | 3626 | |||
XM_017013252.1 | XP_016868741.1 | 18 | 4373 | |||
XM_017013256.1 | XP_016868745.1 | 17 | 3314 | |||
GenBank mRNA | AB018268.1 | - | 15 | 3626 | ||
AK026961.1 | - | 3 | 2037 | |||
AK125904.1 | - | 18 | 4239 | |||
AK127040.1 | - | 9 | 3767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
