siRNA Details
Gene Symbol: SHISA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007538.1 | NP_001007539.1 | 2 | 723 | ||
GenBank mRNA | AY358775.1 | - | 2 | 711 | ||
BC132898.1 | - | 2 | 690 | 40146920 | ||
BC137092.1 | - | 2 | 690 | 9021089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
