siRNA Details
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 24 | 5752 | ||
XM_011511359.1 | XP_011509661.1 | 24 | 5749 | |||
XM_011511361.1 | XP_011509663.1 | 23 | 5506 | |||
XM_011511362.1 | XP_011509664.1 | 23 | 5620 | |||
GenBank mRNA | AB209993.1 | - | 24 | 5670 | ||
AF070582.1 | - | 1 | 663 | |||
AK124683.1 | - | 1 | 1796 | |||
AL050001.1 | - | 1 | 52 | |||
AY726577.1 | - | 1 | 1716 | |||
BC005078.1 | - | 1 | 240 | |||
BC080559.1 | - | 1 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
