siRNA Details
Gene Symbol: FARP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001715.3 | NP_001001715.2 | 2 | 527 | ||
NM_001286839.1 | NP_001273768.1 | 2 | 348 | |||
NM_005766.3 | NP_005757.1 | 2 | 527 | |||
XM_011521046.2 | XP_011519348.1 | 2 | 527 | |||
XM_017020312.1 | XP_016875801.1 | 2 | 271 | |||
GenBank mRNA | AB008430.1 | - | 2 | 157 | ||
BC041595.1 | - | 2 | 374 | 5443230 | ||
BC071592.1 | - | 2 | 348 | 30342239 | ||
BM763139.1 | - | 2 | 271 | |||
CF994038.1 | - | 2 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
