siRNA Details
Gene Symbol: AIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318970.1 | NP_001305899.1 | 5 | 321 | ||
NM_001623.4 | NP_001614.3 | 5 | 382 | |||
NM_032955.2 | NP_116573.1 | 2 | 211 | |||
XM_005248870.4 | XP_005248927.1 | 4 | 572 | |||
XM_017010332.1 | XP_016865821.1 | 4 | 516 | |||
GenBank mRNA | AF088038.1 | - | 5 | 328 | ||
AF299327.1 | - | 2 | 99 | |||
AK290771.1 | - | 5 | 317 | |||
AV716806.1 | - | 5 | 302 | |||
BC009474.1 | - | 5 | 361 | 4249306 | ||
CD249922.1 | - | 2 | 247 | |||
CD521003.1 | - | 5 | 289 | |||
CR542153.1 | - | 2 | 99 | |||
D86438.1 | - | 5 | 270 | |||
EF070981.1 | - | 2 | 104 | |||
EF070982.1 | - | 1 | 319 | |||
EF070983.1 | - | 5 | 261 | |||
JQ782414.1 | - | 5 | 356 | |||
JQ782415.1 | - | 5 | 295 | |||
U19713.1 | - | 5 | 285 | |||
U49392.1 | - | 5 | 346 | |||
U95213.1 | - | 3 | 750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
