siRNA Details
Gene Symbol: AIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318970.1 | NP_001305899.1 | 6 | 588 | ||
NM_001623.4 | NP_001614.3 | 6 | 649 | |||
NM_032955.2 | NP_116573.1 | 3 | 478 | |||
GenBank mRNA | AF088038.1 | - | 6 | 595 | ||
AV716806.1 | - | 6 | 569 | |||
BC009474.1 | - | 6 | 628 | 4249306 | ||
CD249922.1 | - | 3 | 514 | |||
CD521003.1 | - | 6 | 556 | |||
U49392.1 | - | 6 | 607 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
