siRNA Details
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 21 | 3500 | ||
NR_033902.1 | 21 | 3318 | ||||
XM_011544603.1 | XP_011542905.1 | 21 | 3315 | |||
XM_011544604.1 | XP_011542906.1 | 20 | 3198 | |||
XM_017013717.1 | XP_016869206.1 | 21 | 3497 | |||
GenBank mRNA | AB075847.1 | - | 17 | 3091 | ||
AK022661.1 | - | 8 | 1402 | |||
AK095712.1 | - | 21 | 3174 | |||
AK096547.1 | - | 12 | 2149 | |||
AL137523.1 | - | 7 | 1128 | |||
AL834162.1 | - | 21 | 3487 | |||
AL834351.1 | - | 21 | 3488 | |||
AL834352.1 | - | 21 | 3488 | |||
BC018269.1 | - | 9 | 1522 | |||
BC065495.1 | - | 21 | 3176 | 5496068 | ||
BX640952.1 | - | 21 | 3187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
