siRNA Details
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 15, 16 | 2432 | ||
NR_033902.1 | 15, 16 | 2250 | ||||
XM_011544604.1 | XP_011542906.1 | 14, 15 | 2130 | |||
GenBank mRNA | AB075847.1 | - | 12, 13 | 1741 | ||
AF293335.1 | - | 4, 5 | 688 | |||
AK022661.1 | - | 2, 3 | 334 | |||
AK095712.1 | - | 15, 16 | 2106 | |||
AK096547.1 | - | 6, 7 | 1081 | |||
AK314535.1 | - | 15, 16 | 2125 | |||
AL137523.1 | - | 1, 2 | 60 | |||
AL834162.1 | - | 15, 16 | 2419 | |||
AL834351.1 | - | 15, 16 | 2420 | |||
AL834352.1 | - | 15, 16 | 2420 | |||
BC018269.1 | - | 3, 4 | 454 | |||
BC065495.1 | - | 15, 16 | 2108 | 5496068 | ||
BX640952.1 | - | 15, 16 | 2119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
