siRNA Details
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 17 | 2549 | ||
NR_033902.1 | 17 | 2367 | ||||
XM_011544603.1 | XP_011542905.1 | 17 | 2364 | |||
XM_011544604.1 | XP_011542906.1 | 16 | 2247 | |||
XM_017013717.1 | XP_016869206.1 | 17 | 2546 | |||
GenBank mRNA | AB075847.1 | - | 14 | 1858 | ||
AF293335.1 | - | 6 | 805 | |||
AK022661.1 | - | 4 | 451 | |||
AK095712.1 | - | 17 | 2223 | |||
AK096547.1 | - | 8 | 1198 | |||
AK314535.1 | - | 17 | 2242 | |||
AL137523.1 | - | 3 | 177 | |||
AL834162.1 | - | 17 | 2536 | |||
AL834351.1 | - | 17 | 2537 | |||
AL834352.1 | - | 17 | 2537 | |||
BC018269.1 | - | 5 | 571 | |||
BC065495.1 | - | 17 | 2225 | 5496068 | ||
BX640952.1 | - | 17 | 2236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
