siRNA Details
Gene Symbol: OR2T8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005522.1 | NP_001005522.1 | 1 | 433 | ||
XM_011544178.2 | XP_011542480.1 | 2 | 1776 | |||
XM_017001172.1 | XP_016856661.1 | 2 | 4555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
