siRNA Details
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016066.4 | NP_057150.2 | 1, 2 | 603 | ||
GenBank mRNA | AF290514.1 | - | 1, 2 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
