siRNA Details
Gene Symbol: FOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021784.4 | NP_068556.2 | 2 | 1983 | ||
NM_153675.2 | NP_710141.1 | 3 | 1970 | |||
GenBank mRNA | BC006545.2 | - | 2 | 1778 | ||
BC011780.2 | - | 2 | 1824 | 3940139 | ||
BC019288.1 | - | 2 | 1778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
