siRNA Details
Gene Symbol: LINC00167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024233.1 | 1 | 746 | |||
GenBank mRNA | AK055956.1 | - | 1 | 746 | ||
BC137306.1 | - | 1 | 432 | |||
BC137307.1 | - | 1 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
