siRNA Details
Gene Symbol: HIST3H2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033445.2 | NP_254280.1 | 1 | 160 | ||
GenBank mRNA | AK311930.1 | - | 1 | 172 | ||
BC001193.1 | - | 1 | 160 | |||
BC082269.1 | - | 1 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
