siRNA Details
Gene Symbol: ST20-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028330.1 | 1 | 1565 | |||
GenBank mRNA | AK091107.1 | - | 1 | 1565 | ||
AK092679.1 | - | 1 | 1171 | |||
BC016975.1 | - | 1 | 1159 | |||
BG677098.1 | - | 1 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
