siRNA Details
Gene Symbol: ZNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008727.3 | NP_001008727.1 | 4 | 880 | ||
NM_001308269.1 | NP_001295198.1 | 5 | 938 | |||
XM_017027239.1 | XP_016882728.1 | 5 | 1054 | |||
GenBank mRNA | AK297919.1 | - | 5 | 937 | ||
CN360955.1 | - | 1 | 295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
