siRNA Details
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 32 | 7204 | ||
NM_001265611.1 | NP_001252540.1 | 30 | 6355 | |||
NM_014994.2 | NP_055809.2 | 31 | 7186 | |||
NR_049761.1 | 30 | 6996 | ||||
NR_049762.1 | 29 | 6947 | ||||
XM_006720438.1 | XP_006720501.1 | 30 | 7003 | |||
XM_006720439.2 | XP_006720502.1 | 18 | 5278 | |||
XM_011521383.1 | XP_011519685.1 | 31 | 7021 | |||
XM_017022017.1 | XP_016877506.1 | 30 | 7297 | |||
GenBank mRNA | AB011168.2 | - | 32 | 7117 | ||
AL833267.1 | - | 31 | 7113 | |||
AL834413.1 | - | 1 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
