siRNA Details
Gene Symbol: DNMT3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320893.1 | NP_001307822.1 | 18 | 3464 | ||
NM_022552.4 | NP_072046.2 | 23 | 4150 | |||
NM_153759.3 | NP_715640.2 | 19 | 3434 | |||
NM_175629.2 | NP_783328.1 | 23 | 4221 | |||
NR_135490.1 | 24 | 4420 | ||||
XM_005264175.4 | XP_005264232.1 | 23 | 4084 | |||
XM_005264177.4 | XP_005264234.1 | 18 | 3376 | |||
XM_011532662.2 | XP_011530964.1 | 21 | 4086 | |||
XM_011532663.2 | XP_011530965.1 | 24 | 4263 | |||
XM_011532666.2 | XP_011530968.1 | 19 | 3449 | |||
XM_011532667.2 | XP_011530969.1 | 18 | 3266 | |||
XM_017003526.1 | XP_016859015.1 | 23 | 4192 | |||
XM_017003527.1 | XP_016859016.1 | 18 | 3420 | |||
GenBank mRNA | AB208833.1 | - | 24 | 4318 | ||
AF331856.1 | - | 23 | 4100 | |||
BC043617.1 | - | 23 | 4120 | 6150112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
