siRNA Details
Gene Symbol: ATG16L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318766.1 | NP_001305695.1 | 10, 11 | 1259 | ||
NM_033388.1 | NP_203746.1 | 10, 11 | 1139 | |||
XM_005274376.4 | XP_005274433.1 | 10, 11 | 1138 | |||
XM_006718732.2 | XP_006718795.1 | 9, 10 | 1075 | |||
XM_006718733.3 | XP_006718796.1 | 10, 11 | 1091 | |||
XM_006718734.2 | XP_006718797.1 | 2, 3 | 2318 | |||
XM_011545332.1 | XP_011543634.1 | 9, 10 | 1024 | |||
XM_011545333.1 | XP_011543635.1 | 10, 11 | 1587 | |||
XM_011545334.1 | XP_011543636.1 | 10, 11 | 1000 | |||
GenBank mRNA | AK024423.1 | - | 3, 4 | 2876 | ||
AK090597.1 | - | 10, 11 | 1250 | |||
AK091151.1 | - | 9, 10 | 2727 | |||
AK093017.1 | - | 10, 11 | 1112 | |||
AL832974.1 | - | 2, 3 | 750 | |||
BC036713.2 | - | 8, 9 | 868 | |||
BC137489.1 | - | 10, 11 | 1237 | 9021489 | ||
BC137490.1 | - | 10, 11 | 1237 | 9021490 | ||
BC142718.1 | - | 8, 9 | 2835 | |||
BC146660.1 | - | 8, 9 | 2835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
