siRNA Details
Gene Symbol: ATG16L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318766.1 | NP_001305695.1 | 18 | 2197 | ||
NM_033388.1 | NP_203746.1 | 18 | 2077 | |||
XM_006718732.2 | XP_006718795.1 | 17 | 2013 | |||
XM_006718734.2 | XP_006718797.1 | 10 | 3256 | |||
XM_011545332.1 | XP_011543634.1 | 17 | 1962 | |||
XM_011545333.1 | XP_011543635.1 | 18 | 2525 | |||
XM_011545334.1 | XP_011543636.1 | 18 | 1938 | |||
GenBank mRNA | AK024423.1 | - | 8 | 4511 | ||
AK090597.1 | - | 18 | 2188 | |||
AK091151.1 | - | 17 | 3665 | |||
AK093017.1 | - | 18 | 2050 | |||
AL832974.1 | - | 10 | 1688 | |||
BC036713.2 | - | 16 | 1806 | |||
BC142718.1 | - | 16 | 3773 | |||
BC146660.1 | - | 16 | 3773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
