siRNA Details
Gene Symbol: FBXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012175.3 | NP_036307.2 | 5 | 537 | ||
NM_033406.2 | NP_208385.1 | 5 | 537 | |||
XM_011519980.1 | XP_011518282.1 | 5 | 543 | |||
XM_011519981.2 | XP_011518283.1 | 5 | 543 | |||
XM_011519982.1 | XP_011518284.1 | 6 | 597 | |||
XM_017017533.1 | XP_016873022.1 | 6 | 597 | |||
GenBank mRNA | AB209039.1 | - | 4 | 3922 | ||
AF174595.1 | - | 5 | 505 | |||
AF176702.1 | - | 5 | 494 | |||
AK001943.1 | - | 5 | 528 | |||
AK128454.1 | - | 5 | 515 | |||
AL136625.1 | - | 5 | 519 | |||
AL832826.1 | - | 4 | 1209 | |||
BC039291.1 | - | 5 | 537 | |||
BC046110.1 | - | 5 | 523 | 5549548 | ||
BG716664.1 | - | 5 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
