siRNA Details
Gene Symbol: FBXO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012175.3 | NP_036307.2 | 2 | 178 | ||
NM_033406.2 | NP_208385.1 | 2 | 178 | |||
XM_011519980.1 | XP_011518282.1 | 2 | 184 | |||
XM_011519981.2 | XP_011518283.1 | 2 | 184 | |||
XM_011519982.1 | XP_011518284.1 | 2 | 157 | |||
XM_017017533.1 | XP_016873022.1 | 2 | 157 | |||
GenBank mRNA | AB209039.1 | - | 1 | 3563 | ||
AF174595.1 | - | 2 | 138 | |||
AF176702.1 | - | 2 | 135 | |||
AK001943.1 | - | 2 | 169 | |||
AK128454.1 | - | 2 | 156 | |||
AK304866.1 | - | 2 | 173 | |||
AL136625.1 | - | 2 | 161 | |||
AL832826.1 | - | 1 | 850 | |||
BC039291.1 | - | 2 | 178 | |||
BC046110.1 | - | 2 | 164 | 5549548 | ||
BG716664.1 | - | 2 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
