siRNA Details
Gene Symbol: LINC00476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023389.1 | 1 | 759 | |||
GenBank mRNA | AF170307.1 | - | 1 | 262 | ||
AF194971.1 | - | 1 | 758 | |||
AK092137.1 | - | 1 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
