siRNA Details
Gene Symbol: OR6C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005494.1 | NP_001005494.1 | 1 | 139 | ||
GenBank mRNA | BC137000.1 | - | 1 | 167 | 9020992 | |
BC137001.1 | - | 1 | 167 | 9020993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
