siRNA Details
Gene Symbol: VARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167733.2 | NP_001161205.1 | 29 | 3232 | ||
NM_001167734.1 | NP_001161206.1 | 30 | 3467 | |||
NM_020442.5 | NP_065175.4 | 30 | 3460 | |||
GenBank mRNA | AB067472.1 | - | 29 | 3928 | ||
AK000511.1 | - | 29 | 3137 | |||
AK025618.1 | - | 18 | 2052 | |||
AK094483.1 | - | 6 | 1855 | |||
AK125069.1 | - | 19 | 3283 | |||
AK293780.1 | - | 29 | 3221 | |||
AK303415.1 | - | 30 | 3449 | |||
AK304807.1 | - | 30 | 3467 | |||
AL122037.1 | - | 7 | 1056 | |||
BC008844.2 | - | 13 | 1655 | |||
BC009355.2 | - | 20 | 2308 | |||
BC063427.1 | - | 18 | 2083 | |||
BC073838.1 | - | 18 | 2078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
