siRNA Details
Gene Symbol: VARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167733.2 | NP_001161205.1 | 23 | 2197 | ||
NM_001167734.1 | NP_001161206.1 | 24 | 2432 | |||
NM_020442.5 | NP_065175.4 | 24 | 2425 | |||
GenBank mRNA | AB067472.1 | - | 23 | 2893 | ||
AK000511.1 | - | 23 | 2102 | |||
AK025618.1 | - | 12 | 1017 | |||
AK026063.1 | - | 3 | 205 | |||
AK125069.1 | - | 13 | 2248 | |||
AK293780.1 | - | 23 | 2186 | |||
AK303415.1 | - | 24 | 2414 | |||
AK304807.1 | - | 24 | 2432 | |||
AK315733.1 | - | 13 | 1183 | |||
AL122037.1 | - | 1 | 21 | |||
BC008844.2 | - | 7 | 620 | |||
BC009355.2 | - | 14 | 1273 | |||
BC063427.1 | - | 12 | 1048 | |||
BC073838.1 | - | 12 | 1043 | |||
BC112054.1 | - | 23 | 2287 | 8327522 | ||
BC113605.1 | - | 23 | 2287 | 8322657 | ||
BC143534.1 | - | 22 | 2215 | |||
BC143535.1 | - | 23 | 2286 | |||
BC143536.1 | - | 23 | 2280 | 9052045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
