siRNA Details
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF380580.1 | - | 15 | 1667 | ||
AF419616.1 | - | 23 | 2616 | |||
AY894565.1 | - | 19 | 2493 | |||
BX538005.1 | - | 28 | 3928 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 16641 | |||
GenBank mRNA | AK299612.1 | - | 8 | 815 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894568.1 | - | 19 | 2449 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 21 | 2721 | ||
NM_001277444.1 | NP_001264373.1 | 25 | 3228 | |||
XM_006711317.2 | XP_006711380.1 | 26 | 3456 | |||
GenBank mRNA | AF419619.1 | - | 2 | 84 | ||
AK294414.1 | - | 19 | 2386 | |||
AY894574.1 | - | 21 | 2723 | |||
BQ436001.1 | - | 5 | 455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
