siRNA Details
Gene Symbol: TCEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006756.3 | NP_006747.1 | 10 | 2728 | ||
NM_201437.2 | NP_958845.1 | 9 | 2665 | |||
NR_109901.1 | 12 | 3104 | ||||
NR_109902.1 | 5 | 2176 | ||||
XM_006716467.1 | XP_006716530.1 | 11 | 2631 | |||
XM_011517578.1 | XP_011515880.1 | 10 | 2659 | |||
GenBank mRNA | BC072460.1 | - | 10 | 2524 | 6470030 | |
BX538034.1 | - | 12 | 2915 | |||
M81601.1 | - | 10 | 2570 | |||
X57198.1 | - | 9 | 2457 | |||
X62585.1 | - | 10 | 2500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
