siRNA Details
Gene Symbol: FLJ40194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034161.1 | 2 | 284 | |||
GenBank mRNA | AK097513.1 | - | 2 | 288 | ||
BI463223.1 | - | 2 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
