siRNA Details
Gene Symbol: DEFB112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037498.1 | NP_001032587.1 | 2 | 168 | ||
GenBank mRNA | AY122469.1 | - | 1 | 12 | ||
DQ012016.1 | - | 2 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
