siRNA Details
Gene Symbol: DCAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005828.4 | NP_005819.3 | 3 | 598 | ||
NR_073585.1 | 3 | 598 | ||||
GenBank mRNA | AK294925.1 | - | 3 | 572 | ||
BC001264.1 | - | 3 | 551 | 3458293 | ||
BM559804.1 | - | 571 | ||||
BQ221019.1 | - | 3 | 558 | |||
U94747.1 | - | 3 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
