siRNA Details
Gene Symbol: WDR33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018383.4 | NP_060853.3 | 21 | 3783 | ||
XM_005263697.2 | XP_005263754.1 | 20 | 3344 | |||
XM_011511436.1 | XP_011509738.1 | 21 | 3812 | |||
GenBank mRNA | AB044749.1 | - | 21 | 3743 | ||
AK098003.1 | - | 6 | 1245 | |||
AL834365.1 | - | 1 | 28 | |||
BC010283.1 | - | 6 | 1128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
