siRNA Details
Gene Symbol: WDR33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018383.4 | NP_060853.3 | 9 | 1117 | ||
XM_005263697.2 | XP_005263754.1 | 9 | 1117 | |||
XM_011511436.1 | XP_011509738.1 | 9 | 1146 | |||
GenBank mRNA | AB044749.1 | - | 9 | 1077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
