siRNA Details
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 23 | 3557 | ||
XM_011523768.2 | XP_011522070.1 | 22 | 3488 | |||
GenBank mRNA | AB014564.1 | - | 19 | 2946 | ||
AK023003.1 | - | 15 | 2406 | |||
AK074252.1 | - | 8 | 751 | |||
AK125717.1 | - | 23 | 3576 | |||
AK308854.1 | - | 7 | 912 | |||
BC004266.1 | - | 5 | 507 | |||
BC033614.1 | - | 5 | 373 | |||
BC063786.1 | - | 17 | 2639 | |||
BX648864.1 | - | 2 | 376 | |||
CR627449.1 | - | 21 | 3295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
