siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 58 | 9143 | ||
XM_005262858.4 | XP_005262915.1 | 60 | 9184 | |||
XM_011531757.2 | XP_011530059.1 | 59 | 9095 | |||
XM_011531759.2 | XP_011530061.1 | 58 | 9057 | |||
XM_011531760.2 | XP_011530062.1 | 58 | 8995 | |||
XM_011531761.2 | XP_011530063.1 | 59 | 9050 | |||
XM_011531762.2 | XP_011530064.1 | 58 | 9044 | |||
XM_011531763.2 | XP_011530065.1 | 57 | 8941 | |||
XM_011531764.2 | XP_011530066.1 | 58 | 9011 | |||
XM_011531765.2 | XP_011530067.1 | 55 | 8746 | |||
XM_017007906.1 | XP_016863395.1 | 61 | 9312 | |||
XM_017007907.1 | XP_016863396.1 | 61 | 9309 | |||
XM_017007908.1 | XP_016863397.1 | 60 | 9241 | |||
XM_017007909.1 | XP_016863398.1 | 46 | 6937 | |||
GenBank mRNA | AB023210.2 | - | 23 | 2826 | ||
AF538685.1 | - | 57 | 9035 | |||
AK055806.1 | - | 9 | 790 | |||
AK094910.1 | - | 14 | 1544 | |||
BC065502.1 | - | 5 | 493 | |||
BC150246.1 | - | 23 | 2847 | |||
BX648077.1 | - | 15 | 1708 | |||
BX648153.1 | - | 18 | 2036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
