siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 63 | 10033 | ||
XM_005262858.4 | XP_005262915.1 | 65 | 10074 | |||
XM_011531757.2 | XP_011530059.1 | 64 | 9985 | |||
XM_011531759.2 | XP_011530061.1 | 63 | 9947 | |||
XM_011531760.2 | XP_011530062.1 | 63 | 9885 | |||
XM_011531761.2 | XP_011530063.1 | 64 | 9940 | |||
XM_011531762.2 | XP_011530064.1 | 63 | 9934 | |||
XM_011531763.2 | XP_011530065.1 | 62 | 9831 | |||
XM_011531764.2 | XP_011530066.1 | 63 | 9901 | |||
XM_011531765.2 | XP_011530067.1 | 60 | 9636 | |||
XM_017007906.1 | XP_016863395.1 | 66 | 10202 | |||
XM_017007907.1 | XP_016863396.1 | 66 | 10199 | |||
XM_017007908.1 | XP_016863397.1 | 65 | 10131 | |||
XM_017007909.1 | XP_016863398.1 | 51 | 7827 | |||
GenBank mRNA | AB023210.2 | - | 28 | 3716 | ||
AF538685.1 | - | 62 | 9925 | |||
AK055806.1 | - | 14 | 1680 | |||
AK092394.1 | - | 3 | 412 | |||
BC015214.1 | - | 3 | 359 | |||
BC065502.1 | - | 10 | 1383 | |||
BC150246.1 | - | 28 | 3737 | |||
BX648077.1 | - | 20 | 2598 | |||
BX648153.1 | - | 23 | 2926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
