siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 68 | 13176 | ||
XM_005262858.4 | XP_005262915.1 | 70 | 13217 | |||
XM_011531757.2 | XP_011530059.1 | 69 | 13128 | |||
XM_011531759.2 | XP_011530061.1 | 68 | 13090 | |||
XM_011531760.2 | XP_011530062.1 | 68 | 13028 | |||
XM_011531761.2 | XP_011530063.1 | 69 | 13083 | |||
XM_011531762.2 | XP_011530064.1 | 68 | 13077 | |||
XM_011531763.2 | XP_011530065.1 | 67 | 12974 | |||
XM_011531764.2 | XP_011530066.1 | 68 | 13044 | |||
XM_011531765.2 | XP_011530067.1 | 65 | 12779 | |||
XM_017007906.1 | XP_016863395.1 | 71 | 13345 | |||
XM_017007907.1 | XP_016863396.1 | 71 | 13342 | |||
XM_017007908.1 | XP_016863397.1 | 70 | 13274 | |||
XM_017007909.1 | XP_016863398.1 | 56 | 10970 | |||
GenBank mRNA | AB023210.2 | - | 33 | 6859 | ||
AF131845.1 | - | 1 | 762 | |||
BC150246.1 | - | 33 | 6880 | |||
BX648077.1 | - | 25 | 5739 | |||
BX648153.1 | - | 28 | 6071 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
