siRNA Details
Gene Symbol: KCTD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015353.2 | NP_056168.1 | 6 | 1392 | ||
NR_110834.1 | 7 | 1273 | ||||
NR_110835.1 | 8 | 1355 | ||||
GenBank mRNA | BC033329.1 | - | 7 | 1273 | ||
BC080561.1 | - | 1 | 340 | |||
D79998.1 | - | 6 | 1331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
