siRNA Details
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303281.1 | NP_001290210.1 | 2, 3 | 554 | ||
NM_001303282.1 | NP_001290211.1 | 2, 3 | 554 | |||
NM_144680.3 | NP_653281.2 | 4, 5 | 997 | |||
XM_011524002.2 | XP_011522304.1 | 2, 3 | 627 | |||
XM_017025006.1 | XP_016880495.1 | 2, 3 | 554 | |||
XM_017025007.1 | XP_016880496.1 | 3, 4 | 897 | |||
XM_017025008.1 | XP_016880497.1 | 2, 3 | 554 | |||
XM_017025009.1 | XP_016880498.1 | 1, 2 | 882 | |||
XM_017025010.1 | XP_016880499.1 | 2, 3 | 627 | |||
XM_017025011.1 | XP_016880500.1 | 3, 4 | 897 | |||
GenBank mRNA | AK057783.1 | - | 1, 2 | 305 | ||
AK092656.1 | - | 4, 5 | 997 | |||
AK127908.1 | - | 2, 3 | 512 | |||
AY491053.1 | - | 2, 3 | 501 | |||
BC036096.2 | - | 1, 2 | 475 | 5301103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
