siRNA Details
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303281.1 | NP_001290210.1 | 2 | 296 | ||
NM_001303282.1 | NP_001290211.1 | 2 | 296 | |||
NM_144680.3 | NP_653281.2 | 4 | 739 | |||
XM_011524002.2 | XP_011522304.1 | 2 | 369 | |||
XM_017025006.1 | XP_016880495.1 | 2 | 296 | |||
XM_017025007.1 | XP_016880496.1 | 3 | 639 | |||
XM_017025008.1 | XP_016880497.1 | 2 | 296 | |||
XM_017025009.1 | XP_016880498.1 | 1 | 624 | |||
XM_017025010.1 | XP_016880499.1 | 2 | 369 | |||
XM_017025011.1 | XP_016880500.1 | 3 | 639 | |||
GenBank mRNA | AK057783.1 | - | 1 | 47 | ||
AK092656.1 | - | 4 | 739 | |||
AK127908.1 | - | 2 | 254 | |||
AY491053.1 | - | 2 | 243 | |||
BC036096.2 | - | 1 | 217 | 5301103 | ||
DB056532.1 | - | 2 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
