siRNA Details
Gene Symbol: MINOS1-NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204088.1 | NP_001191017.1 | 1 | 100 | ||
NM_001204089.1 | NP_001191018.1 | 1 | 100 | |||
GenBank mRNA | AK289456.1 | - | 1 | 98 |
Gene Symbol: MINOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032363.3 | NP_001027535.1 | 1 | 100 | ||
NM_001204082.1 | NP_001191011.1 | 1 | 100 | |||
NM_001204083.1 | NP_001191012.1 | 1 | 100 | |||
NR_033757.2 | 1 | 100 | ||||
NR_033758.2 | 1 | 100 | ||||
GenBank mRNA | AJ711911.1 | - | 1 | 97 | ||
AK094318.1 | - | 1 | 77 | |||
BC009927.1 | - | 1 | 76 | |||
BC070388.1 | - | 1 | 87 | 30407082 | ||
BG687187.1 | - | 108 | ||||
BM784596.1 | - | 1 | 53 | |||
CB157536.1 | - | 1 | 36 | |||
DA523181.1 | - | 1 | 100 | |||
DA793500.1 | - | 1 | 57 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
