siRNA Details
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008224.1 | NP_001008225.1 | 16 | 2771 | ||
NM_018003.2 | NP_060473.2 | 16 | 2548 | |||
XM_005254529.4 | XP_005254586.1 | 15 | 2765 | |||
XM_011521752.2 | XP_011520054.2 | 16 | 2877 | |||
XM_011521753.2 | XP_011520055.2 | 15 | 2845 | |||
XM_011521754.2 | XP_011520056.1 | 15 | 2326 | |||
XM_017022394.1 | XP_016877883.1 | 15 | 2597 | |||
XM_017022395.1 | XP_016877884.1 | 16 | 2408 | |||
GenBank mRNA | AB046781.2 | - | 16 | 2444 | ||
AF322916.1 | - | 16 | 2493 | |||
AK000990.1 | - | 1 | 509 | |||
BC113407.1 | - | 16 | 2467 | 8322459 | ||
BC113409.1 | - | 16 | 2467 | 8322461 | ||
BC143266.1 | - | 16 | 2467 | 9051768 | ||
BC143267.1 | - | 13 | 2140 | 9051769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
