siRNA Details
Gene Symbol: CDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004361.3 | NP_004352.2 | 12 | 3059 | ||
NM_033646.2 | NP_387450.1 | 12 | 2689 | |||
XM_017025523.1 | XP_016881012.1 | 12 | 3375 | |||
GenBank mRNA | AB035301.1 | - | 12 | 2790 | ||
AJ007611.1 | - | 12 | 2684 | |||
CN309979.1 | - | 1 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
