siRNA Details
Gene Symbol: CDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317214.1 | NP_001304143.1 | 10 | 2262 | ||
NM_004361.3 | NP_004352.2 | 10 | 2262 | |||
NM_033646.2 | NP_387450.1 | 10 | 1892 | |||
XM_017025523.1 | XP_016881012.1 | 10 | 2578 | |||
GenBank mRNA | AB035301.1 | - | 10 | 1993 | ||
AJ007611.1 | - | 10 | 1890 | |||
BC036786.1 | - | 10 | 2262 | 5744842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
