siRNA Details
Gene Symbol: CDH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317214.1 | NP_001304143.1 | 5, 6 | 1496 | ||
NM_004361.3 | NP_004352.2 | 5, 6 | 1496 | |||
NM_033646.2 | NP_387450.1 | 5, 6 | 1126 | |||
XM_017025523.1 | XP_016881012.1 | 5, 6 | 1812 | |||
GenBank mRNA | AB035301.1 | - | 5, 6 | 1227 | ||
AJ007611.1 | - | 5, 6 | 1124 | |||
BC036786.1 | - | 5, 6 | 1496 | 5744842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
