siRNA Details
Gene Symbol: OR3A4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024128.1 | 1 | 325 | |||
GenBank mRNA | BC136874.1 | - | 1 | 325 | ||
BC136875.1 | - | 1 | 325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
