siRNA Details
Gene Symbol: SLC25A5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028443.1 | 1 | 619 | |||
GenBank mRNA | AK092094.1 | - | 1 | 595 | ||
BC050320.1 | - | 1 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
