siRNA Details
Gene Symbol: GAS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130831.1 | NP_001124303.1 | 14 | 3628 | ||
NM_003644.2 | NP_003635.2 | 10 | 3377 | |||
NM_201432.1 | NP_958836.1 | 14 | 3785 | |||
NM_201433.1 | NP_958839.1 | 14 | 3815 | |||
XM_005256831.3 | XP_005256888.1 | 15 | 3726 | |||
XM_005256832.3 | XP_005256889.1 | 14 | 3596 | |||
XM_005256833.3 | XP_005256890.1 | 13 | 3703 | |||
XM_005256834.3 | XP_005256891.1 | 13 | 3529 | |||
XM_005256835.3 | XP_005256892.1 | 13 | 3473 | |||
XM_011524044.2 | XP_011522346.1 | 14 | 3620 | |||
XM_017025239.1 | XP_016880728.1 | 14 | 3649 | |||
XM_017025240.1 | XP_016880729.1 | 13 | 3432 | |||
XM_017025241.1 | XP_016880730.1 | 13 | 3408 | |||
XM_017025242.1 | XP_016880731.1 | 13 | 3582 | |||
GenBank mRNA | AB007854.1 | - | 14 | 3583 | ||
AY327406.1 | - | 14 | 3815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
